HEK-Blue™ Selection, InvivoGen
Supplier: Invivogen
HB-SEL
MSPP-HB-SELEA 247.69 USD
MSPP-HB-SEL
HEK-Blue™ Selection, InvivoGen
Assays Cellular Assays
HEK-Blue™ Selection is a solution that combines several selective antibiotics. These antibiotics guarantee the persistent expression of the various transgenes introduced in HEK-Blue™ TLR2 and HEK-Blue™ TLR4 cells.
HEK-Blue™ Selection should be used with the following cell lines: HEK-Blue™-2 cells, HEK-Blue™-4 cells, HEK-Blue™ hTLR2 cells, HEK-Blue™ mTLR2 cells, HEK-Blue™ hTLR4 cells, HEK-Blue™ mTLR4 cells, HEK-Blue™ IL-12 cells, HEK-Blue™ IL-17 cells and HEK-Blue™ IL-33 cells.