Goldenrod Animal Lancet, Braintree Scientific
Supplier: 0000018294
GR-4MM GR-5MM GR-6MM
MSPP-GR4MMCS 95.38 USD
MSPP-GR4MM MSPP-GR5MM MSPP-GR-6MM
Goldenrod Animal Lancet, Braintree Scientific
Blood Lancets
Animal lancet for blood collection of facial vein (submandibular) blood samples.
  • Sterile, disposable blood lancet
  • Inexpensive, easy, and fast to use
Multiple blood samples can be taken from small animals without hurting the animals.
Packaging: Packed in 5 trays of 200.