Anti-KDR Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 89106]

Supplier: R&D Systems
FAB357C-025
MSPP-FAB357C02EA 229.94 USD
MSPP-FAB357C02
Anti-KDR Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 89106]
Antibodies
Type: Primary
Antigen: KDR
Clonality: Monoclonal
Clone: 89106
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR