PBS3 Vertical-Wheel® MagDrive Bioreactor System, PBS Biotech
Supplier: PBS BIOTECH, INC. MS
FA-3-D-706-L
MSPP-FA-3D706LEA 7538.46 USD
MSPP-FA-3D706L
PBS3 Vertical-Wheel® MagDrive Bioreactor System, PBS Biotech
Bioreactors
MagDrive bioreactor system is optimal for culturing shear sensitive cells, such as MSCs or human primary cells grown on microcarriers or hPSCs grown as cell aggregates.
  • Efficient mixing with homogeneous particle suspension and low shear stress
  • Eliminates need for anti-foaming agents or shear protectants
  • Customizable, secure, remote access control and alarm reporting
  • Fully-integrated touchscreen control with network connectivity capability
  • Compact design and small footprint, with simple setup and training requirements: plug-and-play right out of the box
MagDrive bioreactor system can be used for process development that will have predictive performance in progressively larger Vertical-Wheel bioreactors, up to commercial scale​. Magnetic coupling between Vertical-Wheel and housing unit controls agitation speed. Adjustable height dip tube allows for rapid and efficient medium exchange and cell harvesting.
Peristaltic pumps media addition and harvest: fixed-speed/variable-speed uni-directional. Sampling: fixed-speed, bi-directional. Temperature control range can be 5 °C above ambient to 40 °C (±0.2 °C).
Certifications: USP Class VI Testing for Plastics <88> and/or ISO 10993 complaint.