Vacuum/Pressure Pumps Coated with PTFE
Supplier: Cole Parmer
EW-79200-35
MSPP-EW7920035EA 566.51 USD
MSPP-EW7920035
Vacuum/Pressure Pumps Coated with PTFE
Pumps Vacuum Pumps
Wetted parts are coated with PTFE to handle your toughest applications.
  • Lightweight aluminum construction and compact size
  • Pumps with gauges and regulators give you more control of your vacuum/pressure applications
These pumps are ideal for benchtop applications such as vacuum filtration. Pumps include suction cup feet and a 6-ft power cord with built-in switch (the 115 VAC models have grounded plug, 220 VAC and DC models have stripped ends).
Ordering information: Three years warranty.