PCR Micro Reaction Tube, Cole-Parmer
Supplier: Cole Parmer
67104-60
MSPP-EW6710460PK 123.37 USD
MSPP-EW6710460
PCR Micro Reaction Tube, Cole-Parmer
Tubes PCR Tubes
Designed to amplify nucleic acids during the PCR process.
  • Precision molded from 99.9% pure virgin polypropylene - withstands temperatures of –20 to 122 °C and are autoclavable
  • Ultra-thin walls accelerate heat transfer providing shorter cycle times
  • Full-length, even insertion cap ensures a tight fit, eliminating sample evaporation
Certified RNase, DNase, and pyrogen safe to ensure no degradation of the target or amplified product.