Keck Sieve Shaker Kit, Cole Parmer
Supplier: Cole Parmer
59978-10
MSPP-EW5997810EA 420 USD
MSPP-EW5997810
Keck Sieve Shaker Kit, Cole Parmer
Shakers and Mixers Sieve Shakers
Lightweight, compact design, easy to take into the field.
  • Kit includes 20 interchangeable screens in a wide range of mesh sizes
  • Provides clear indication of sample
  • A complete kit with proven, reliable results
This complete mechanical sieve kit provides reliable particle size analysis. Kit includes 20 stainless steel mesh screens and five interlocking cylinders. Shakers are easy to use; place the sample in the top cylinder and shake to distribute particles, the volumetric percentages are indicated on the shaker frame. The clear acrylic cylinders makeMore Product Information