Analog Rollers Shaker, Fixed Speed, RS-200-L-120
Supplier: Cole Parmer
EW-51901-21
MSPP-EW5190121EA 1458.15 USD
MSPP-EW5190121
Analog Rollers Shaker, Fixed Speed, RS-200-L-120
Shakers and Mixers Rotating/Rocking Shakers
Solidly constructed roller shaker is ideal for mixing blood samples, viscous substances, and liquid-solid suspensions where minimum aeration is required.
  • Gentle rocking and rolling action for complete mixing
  • Continuous and quiet operation
  • Side panels ensure sample bottles stay on the shaker
Safe for use in cold rooms down to 4 °C (39 °F) or in incubators to 40 °C (104 °F), up to 80% humidity.
Ordering information: Three year warranty.