Supplier: Cole Parmer
EW-41720-40
MSPP-EW4172040EA 1666.49 USD
MSPP-EW4172040
Lights Halogen Lights, Lamps and Bulbs
Avoid lighting problems due to variation in input voltage.
  • Ideal for video microscopy and machine vision applications
  • One year warranty
The internal power supplyin the illuminator to the halogen bulb is stabilized to ±0.5%, eliminating problems due to variation in input line voltages. Front-panel dial controls light output from 0 to 100%. Remotely control the light output using a 0 to 5 VDC signal. Stackable housing is heavy-duty steel. Includes a 150 W bulb and 6' power cord with three-prong plug.
Illuminator dimension: 18.41W×24.13D×11.75H cm (7.25×9.5×4.6")
Certifications: UL/cUL listed, CE approved, and RoHS compliant.