Heavy-Duty Three-Prong Utility Clamps, Cole-Parmer
Supplier: Cole Parmer
802116
MSPP-EW0802116EA 81.51 USD
MSPP-EW0802116
Heavy-Duty Three-Prong Utility Clamps, Cole-Parmer
Clamps General Purpose Clamps
Securely hold your flasks, beakers and burettes.
  • Supplied with non-slip vinyl sleeves or for temperatures above 212 °F (100 °C), fiberglass covers
  • Available in 3 sizes, small, medium, and large
  • Constructed of nickel plated zinc
Featuring an innovative closed yoke design that minimizes contamination and corrosion of internal components and is easier to clean and decontaminate. This design allows use of the clamp’s full depth – no springs or internal workings to get in the way.
Choose single adjustment style for standard use where one prong or “finger” is adjusted using a wing nut, while the other prong remains stationary or for a more versatile clamp choose the dual adjustment style. Utilizing thumbscrews, both prongs are easily adjusted over a wider range.
The prong height for these clamps make them ideal for use in holding and securing glassware with precision ground glass joints.
Ordering information: An additional holder must be purchased in order to attach clamps to lab-frames, support stands or other apparatus.