Infrared Heating Lamp, Cole Parmer
Supplier: Cole Parmer
03057-00
MSPP-EW0305700EA 567.69 USD
MSPP-EW0305700
Infrared Heating Lamp, Cole Parmer
Lights Infrared Lights, Lamps and Bulbs
Lamp can be mount to any table or benchtop, or use our floor stands to support the heating lamp.
  • Features durable all-metal construction
  • Dual reflector prevents excessive heat buildup
Position of the lamp can be adjusted using 42" swiveling extension arm. Lamp comes complete with screw-type mounting bracket, 10-ft grounded cord with plug and 250 watt, hard glass infrared heat bulb. Easily remove wire bulb guard to replace bulb.
Ordering information: Replacement bulb should be ordered separately.