Test Tube Support System, PP, Cole Parmer
Supplier: Cole Parmer
06733-00
MSPP-EW-067330EA 61.11 USD
MSPP-EW-067330
Test Tube Support System, PP, Cole Parmer
Racks Tube Racks
Rotate each cube independently-use several sizes of test tubes, culture tubes, or microcentrifuge tubes at the same time.
  • Cube modules are color coded for convenience
  • Autoclavable
Compatible with the tubes: 15 to 50 ml conical; 12×75 mm, 13×100 mm, 16×125 mm, and 17×100 mm culture; and 1.5 and 2.0 ml microcentrifuge.
Delivery information: Each system consists of four cubes and one support tray.