Multiple Species Recombinant MT (from Synthetic), BSA (Bovine Serum Albumin)
Supplier: CLOUD-CLONE CORP MS
CPA908GE11
MSPP-CPA908GE1EA 350 USD
MSPP-CPA908GE1
Multiple Species Recombinant MT (from Synthetic), BSA (Bovine Serum Albumin)
Proteins and Peptides
This is a MT conjugated BSA, General species is sequencing from C13H16N2O2 with 90 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Melatonin, 5-methoxy-N-acetyltryptamine, is a hormone found in all living creatures from algae to humans, at levels that vary in a daily cycle. Many biological effects of melatonin are produced through activation of melatonin receptors,while others are due to its role as a pervasive and extremely powerful antioxidant with a particular role in the protection of nuclear and mitochondrial DNA. Melatonin is secreted by the pineal gland in the brain and is important in the regulation of many hormones in the body. Among its key roles, melatonin controls the body's circadian rhythm, an internal 24-hour time-keeping system that plays an important role in when fall asleep and when wake up. Darkness stimulates the release of melatonin and light suppresses its activity.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.