Multiple Species Recombinant Cor (from Synthetic), BSA (Bovine Serum Albumin)
Supplier: CLOUD-CLONE CORP MS
CPA067GE11
MSPP-CPA067GE1EA 380 USD
MSPP-CPA067GE1
Multiple Species Recombinant Cor (from Synthetic), BSA (Bovine Serum Albumin)
Proteins and Peptides
This is a Cor conjugated BSA, General species is sequencing from C21H28O5 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Cortisone is a steroid hormone. Chemically, it is a corticosteroid closely related to corticosterone. It is used to treat a variety of ailments and can be administered intravenously, orally, intraarticularly, or cutaneously. Cortisone suppresses the immune system, thus reducing inflammation and attendant pain and swelling at the site of the injury. However, its effects on the immune system have resulted in various side-effects, particularly in the long-term use of cortisone. 17-hydroxycorticosteroid (17-OHCS) is an inactive product formed when the liver and other body tissues break down cortisol, a steroid hormone.17-OHCS is a group of hydroxycorticosteroids bearing a hydroxy group at the 17-position. Urinary excretion of these compounds is used as an index of adrenal function. They are used systemically in the free alcohol form, but with esterification of the hydroxy groups, topical effectiveness is increased.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.