General Species FA ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEX126GE
MSPP-CEX126GEEA 898 USD
MSPP-CEX126GE
General Species FA ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species FA (Folinic Acid). The assay range is from 117.2 to 30,000 pg/ml (Competitive kit) with a sensitivity of 41.5 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Folinic acid is an adjuvant used in cancer chemotherapy involving the drug methotrexate. It is also used in synergistic combination with the chemotherapy agent 5-fluorouracil. Levofolinic acid and its salts are the enantiopure drugs (in this case, the levo form), and are the only molecules that are biologically active. Folinic acid is a 5-formyl derivative of tetrahydrofolic acid. It is readily converted to other reduced folic acid derivatives, and, thus, has vitamin activity that is equivalent to that of folic acid. Since it does not require the action of dihydrofolate reductase for its conversion, its function as a vitamin is unaffected by inhibition of this enzyme by drugs such as methotrexate. This is the classical view of Folinic acid rescue therapy. Folinic acid is administered at the appropriate time following methotrexate as part of a total chemotherapeutic plan, where it may 'rescue' bone marrow and gastrointestinal mucosa cells from methotrexate.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.