General Species Gly ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CES117GE
MSPP-CES117GEEA 870 USD
MSPP-CES117GE
General Species Gly ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species Gly (Glycine). The assay range is from 1.23 to 100 µg/ml (Competitive kit) with a sensitivity of 0.59 µg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Glycine is an organic compound with the formula NH₂CH₂COOH. With only two hydrogen atoms as its 'side chain', glycine is the smallest of the 20 amino acids commonly found in proteins. Its codons are GGU, GGC, GGA, GGG. Glycine is a colorless, sweet-tasting crystalline solid. It is unique among the proteinogenic amino acids in that it is not chiral. It can fit into hydrophilic or hydrophobic environments, due to its two hydrogen atom side chain. Glycine is an intermediate in the synthesis of a variety of chemical products. It is used in the manufacture of the herbicide glyphosate. Glyphosate is a non-selective systemic herbicide used to kill weeds, especially perennials and broadcast or used in the cut-stump treatment as a forestry herbicide. Initially, glyphosate was sold only by Monsanto under the tradename Roundup, but is no longer under patent.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.