General Species 2-AG ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEO443GE
MSPP-CEO443GEEA 898 USD
MSPP-CEO443GE
General Species 2-AG ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species 2 to AG (2 to Arachidonoylglycerol). The assay range is from 3.70 to 300 ng/ml (Competitive kit) with a sensitivity of 1.44 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
2-Arachidonoylglycerol (2-AG) is an endocannabinoid, an endogenous agonist of the CB1 receptor. It is an ester formed from the omega-6 fatty acid arachidonic acid and glycerol. It is present at relatively high levels in the central nervous system, with cannabinoid neuromodulatory effects. It has been found in maternal bovine and human milk. 2-Arachidonoylglycerol is synthesized from arachidonic acid-containing diacylglycerol (DAG), which is derived from the increase of inositol phospholipid metabolism by the action of diacylglycerol lipase. The molecule can also be formed from pathways like the hydrolysis derived (by diglyceride) from both phosphatidylcholine (PC) and phosphatidic acid (PAs) by the action of DAG lipase and the hydrolysis of arachidonic acid-containing lysophosphatidic acid by the action of a phosphatase.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.