General Species THB ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEG421GE
MSPP-CEG421GEEA 900 USD
MSPP-CEG421GE
General Species THB ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species THB (Tetrahydrobiopterin). The assay range is from 391 to 100000 pg/ml (Competitive kit) with a sensitivity of 137 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Tetrahydrobiopterinis a naturally occurring nutrient and essential cofactor of the three aromatic amino acid hydroxylase enzymes, used in the biosynthesis of the neurotransmitters serotonin (5-hydroxytryptamine (5-HT)), melatonin, dopamine, norepinephrine (noradrenaline), epinephrine (adrenaline), and nitric oxide (NO). A deficit in THB biosynthesis and/or regeneration results in phenylketonuria (PKU) from excess L-phenylalanine concentrations or hyperphenylalaninemia (HPA), as well as monoamine and nitric oxide neurotransmitter deficiency or chemical imbalance. The chronic presence of PKU can result in severe brain damage, including symptoms of mental retardation, microcephaly, speech impediments such as stuttering, slurring, and lisps, seizures or convulsions, and behavioral abnormalities, among other effects.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.