General Species HCy ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CED984GE
MSPP-CED984GEEA 816 USD
MSPP-CED984GE
General Species HCy ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species HCy (Homocysteine). The assay range is from 98.77 to 8000 ng/ml (Competitive kit) with a sensitivity of 40.22 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Homocysteine is an amino acid with the formula HSCH₂CH₂CH(NH₂)CO₂H. It is a homologue of the amino acid cysteine, differing by an additional methylene (-CH₂-) group. It is biosynthesized from methionine by the removal of its terminal Cε methyl group. Homocysteine can be recycled into methionine or converted into cysteine with the aid of B-vitamins. Homocysteine is not obtained from the diet. Instead, it is biosynthesized from methionine via a multi-step process. First, methionine receives an adenosine group from ATP, a reaction catalyzed by S-Adenosyl-methionine synthetase, to give S-adenosyl methionine ('SAM'). SAM then transfers the methyl group to an acceptor molecule. The adenosine is then hydrolyzed to yield L-homocysteine. L-homocysteine has two primary fates: conversion via tetrahydrofolate (THF) back into L-methionine or conversion to L-cysteine.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.