Human CAMP ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEC419HU
MSPP-CEC419HUEA 680 USD
MSPP-CEC419HU
Human CAMP ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human CAMP (Cathelicidin Antimicrobial Peptide). The assay range is from 123.5 to 10,000 ng/ml (Competitive kit) with a sensitivity of 49.4 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Cathelicidin antimicrobial peptide is a family of polypeptides found in lysosomes in polymorphonuclear leukocytes (PMNs). Members of the cathelicidin family of antimicrobial polypeptides are characterized by a highly conserved region (cathelin domain) and a highly variable cathelicidin peptide domain. Cathelicidin peptides have been isolated from many different species of mammals. Cathelicidins were originally found in neutrophils but have since been found in many other cells including epithelial cells and macrophages activated by bacteria, viruses, fungi, or the hormone 1,25-D. The cathelicidin family shares primary sequence homology with the cathepsin family of cysteine proteinase inhibitors, although amino acid residues thought to be important in such protease inhibition are usually lacking.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.