Human AP13 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEB887HU
MSPP-CEB887HUEA 665 USD
MSPP-CEB887HU
Human AP13 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human AP13 (Apelin 13). The assay range is from 12.35 to 1000 pg/ml (Competitive kit) with a sensitivity of 4.83 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Apelin is the endogenous ligand for the G-protein-coupled APJ receptor that is expressed at the surface of some cell types. It is widely expressed in various organs such as the heart, lung, kidney, liver, adipose tissue,gastrointestinal tract, brain, adrenal glands, endothelium, and human plasma. Apelin gene encodes a pre-proprotein of 77 amino acids, with a signal peptide in the N-terminal region. After translocation into the endoplasmic reticulum and cleavage of the signal peptide, the proprotein of 55 amino acids may generate several active fragments: a 36 amino acid peptide corresponding to the sequence 42-77 (apelin 36), a 17 amino acid peptide corresponding to the sequence 61-77 (apelin 17) and a 13 amino acid peptide corresponding to the sequence 65-77 (apelin 13). This later fragment may also undergo a pyroglutamynation at the level of its N-terminal glutamine residue.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.