Rat GT ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEB224RA
MSPP-CEB224RAEA 722 USD
MSPP-CEB224RA
Rat GT ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat GT (Gastrin). The assay range is from 12.35 to 1000 pg/ml (Competitive kit) with a sensitivity of 4.54 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Gastrin is a major physiological regulator of gastric acid secretion. It also has an important trophic or growth-promoting influence on the gastric mucosa. Gastrin is synthesized in G cells, which are located in gastric pits, primarily in the antrum region of the stomach and binds receptors found predominantly on parietal and enterochromaffin-like cells.In humans, gastrin is a hormone that stimulates secretion of gastric acid by the parietal cells of the stomach. It is released by G cells in the stomach and duodenum.Gastrin is a linear peptide hormone produced by G cells of the duodenum and in the pyloric antrum of the stomach. In the Zollinger-Ellison syndrome, gastrin is produced at excessive levels, often by a gastrinoma of the antrum or the pancreas.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.