Human GT ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEB224HU
MSPP-CEB224HUEA 665 USD
MSPP-CEB224HU
Human GT ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human GT (Gastrin). The assay range is from 12.35 to 1000 pg/ml (Competitive kit) with a sensitivity of 4.89 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Gastrin is a major physiological regulator of gastric acid secretion. It also has an important trophic or growth-promoting influence on the gastric mucosa. Gastrin is synthesized in G cells, which are located in gastric pits, primarily in the antrum region of the stomach and binds receptors found predominantly on parietal and enterochromaffin-like cells. In humans, gastrin is a hormone that stimulates secretion of gastric acid by the parietal cells of the stomach. It is released by G cells in the stomach and duodenum. Gastrin is a linear peptide hormone produced by G cells of the duodenum and in the pyloric antrum of the stomach. In the Zollinger-Ellison syndrome, gastrin is produced at excessive levels, often by a gastrinoma of the antrum or the pancreas.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.