Human AOPP ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEB223HU
MSPP-CEB223HUEA 665 USD
MSPP-CEB223HU
Human AOPP ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human AOPP (Advanced Oxidation Protein Products). The assay range is from 61.7 to 5000 ng/ml (Competitive kit) with a sensitivity of 25.3 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
AOPP are carried by oxidized plasma proteins, especially albumin and do not have oxidant properties. AOPP increased in a dose-dependent manner following in vitro exposure of plasma or purified human serum albumin (HSA) to hypochlorous acid. Advanced glycation end products of human serum albumin (AGE-HSA) also increased AOPP levels. in vivo, plasma level of AOPP was the highest in patients on hemodialysis, followed by those on peritoneal dialysis and by undialyzed patients with advanced chronic renal failure. AOPP levels correlated with plasma concentrations of dityrosine and AGE-pentosidine, as indices of oxidant-mediated protein damage, but not with thiobarbituric reactive substances as lipid peroxidation markers. A close correlation was also found between AOPP and neopterin levels, suggesting that AOPP could be part in the monocyte-mediated inflammatory disorders associated with uremia.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.