Human NT ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEB203HU
MSPP-CEB203HUEA 665 USD
MSPP-CEB203HU
Human NT ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human NT (Neurotensin). The assay range is from 37.0 to 3000 pg/ml (Competitive kit) with a sensitivity of 13.2 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Neurotensin is a 13 amino acid neuropeptide that is implicated in the regulation of luteinizing hormone and prolactin release and has significant interaction with the dopaminergic system. Neurotensin was first isolated from extracts of bovine hypothalamus based on its ability to cause a visible vasodilation in the exposed cutaneous regions of anesthetized rats. The sequence of bovine neurotensin was determined to be pyroGlu-Leu-Tyr-Glu-Asn-Lys-Pro-Arg-Arg-Pro-Tyr-Ile-Leu-OH. Neurotensin is synthesized as part of a 169 to 170 amino acid precursor protein that also contains the related neuropeptide neuromedin N. The peptide coding domains are located in tandem near the carboxyl terminal end of the precursor and are bounded and separated by paired basic amino acid (lysine-arginine) processing sites.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.