General Species ADP ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEB069GE
MSPP-CEB069GEEA 870 USD
MSPP-CEB069GE
General Species ADP ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species ADP (Adenosine Diphosphate). The assay range is from 18.52 to 1,500 ng/ml (Competitive kit) with a sensitivity of 7.66 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Adenosine diphosphate, abbreviated ADP, is a nucleotide. It is an ester of pyrophosphoric acid with the nucleoside adenosine. ADP consists of the pyrophosphate group, the pentose sugar ribose, and the nucleobase adenine. ADP is the product of ATP dephosphorylation by ATPases. ADP is converted back to ATP by ATP synthases. ATP is an important energy transfer molecule in cells. ADP is stored in dense bodies inside blood platelets and is released upon platelet activation. ADP interacts with a family of ADP receptors found on platelets (P2Y1, P2Y12 and P2X1), leading to further platelet activation. ADP in the blood is converted to adenosine by the action of ecto-ADPases, inhibiting further platelet activation via adenosine receptors. The anti-platelet drug Plavix (clopidogrel) inhibits the P2Y12 receptor.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.