General Species 1,5-AG ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEB046GE
MSPP-CEB046GEEA 808 USD
MSPP-CEB046GE
General Species 1,5-AG ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species 1,5 to AG (1,5 to Anhydroglucitol). The assay range is from 1.9 to 150ug/mL (Competitive kit) with a sensitivity of 0.7ug/mL. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
1,5-Anhydroglucitol, also known as 1,5-AG, is used primarily to identify glycemic variability in people with diabetes who have normal or near normal hemoglobin A1c levels. 1,5-Anhydroglucitol is a naturally occurring monosaccharide found in nearly all foods, and once ingested is almost 100% non metabolized. 1,5-Anhydroglucitol values decrease during times of hyperglycemia above 180 mg/dL, and return to normal levels after approximately 2 weeks in the absence of hyperglycemia. Monitoring 1,5-anhydroglucitol in people with either type-1 or type-2 diabetes mellitus can assist diabetic patients in determining if their blood glucose levels are frequently above 180 mg/dL, even in the presence of relatively good hemoglobin A1c levels or blood glucose monitoring.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.