General Species ALD ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA911GE
MSPP-CEA911GEEA 796 USD
MSPP-CEA911GE
General Species ALD ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species ALD (Aldosterone). The assay range is from 24.69 to 2,000 pg/ml (Competitive kit) with a sensitivity of 9.41 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Aldosterone is a steroid hormone with a molecular weight of 360.4. It is secreted by the glomerulosa zone of the adrenal gland and controls regulation of hydromineral metabolism. In the distal part of the nephron, aldosterone encourages reabsorption of Na and Cl- ions and secretion of K and H ions in blood vessel lumen. Thus there is an increase in extra-cellular osmolarity and water retention. Regulation of aldosterone secretion is linked to the body's water balance and brings three factors into play. These are the renin- angiotensin system, the plasmic Na/K ratio and, as a secondary factor, the ACTH. Physiologically, there seems to be circadian rythm for levels of aldosterone in the blood, which may be linked to changes in body posture. Aldosterone levels are higher for children than for adults, and increase during pregnancy.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.