General Species NE ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA907GE
MSPP-CEA907GEEA 796 USD
MSPP-CEA907GE
General Species NE ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species NE (Noradrenaline). The assay range is from 61.7 to 5,000 pg/ml (Competitive kit) with a sensitivity of 24.4 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Noradrenaline is the neurotransmitter of the peripheral sympathetic nervous system, responsible for so-called adrenergic neurotransmission, at nerve endings in the heart, in smooth (involuntary) muscle, and in many glands. It is one of the catecholamines. Tyrosine, an amino acid present in body fluids, is taken up into the adrenergic nerve terminal where it is acted upon by an enzyme, tyrosine hydroxylase, to form DOPA, this is converted to dopamine and in turn to noradrenaline. The neurotransmitter is stored in small vesicles, awaiting release. Arrival of the impulse at the nerve terminal releases vesicles, freeing the noradrenaline to exert its effects. To terminate the action of the transmitter, some noradrenaline is oxidized to inactive material, but most is taken back up into the nerve terminal and stored for later use. There are two types of cell membrane receptors with which noradrenaline interacts, termed a and b, each group having several subtypes.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.