Rat NPY ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA879RA
MSPP-CEA879RAEA 684 USD
MSPP-CEA879RA
Rat NPY ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat NPY (Neuropeptide Y). The assay range is from 2.47 to 200 pg/ml (Competitive kit) with a sensitivity of 0.94 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Neuropeptide Y (NPY) is a 36 amino acid peptide neurotransmitter found in the brain and autonomic nervous system. NPY is a neuropeptide that is widely expressed in the central nervous system and influences many physiological processes, including cortical excitability, stress response, food intake, circadian rhythms, and cardiovascular function. The neuropeptide functions through G protein-coupled receptors to inhibit adenylyl cyclase, activate mitogen-activated protein kinase (MAPK), regulate intracellular calcium levels, and activate potassium channels. A polymorphism in this gene resulting in a change of leucine 7 to proline in the signal peptide is associated with elevated cholesterol levels, higher alcohol consumption, and may be a risk factor for various metabolic and cardiovascular diseases.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.