Dog ACTH ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA836CA
MSPP-CEA836CAEA 720 USD
MSPP-CEA836CA
Dog ACTH ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Dog ACTH (Adrenocorticotropic Hormone). The assay range is from 12.35 to 1000 pg/ml (Competitive kit) with a sensitivity of 5.06 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
ACTH (Adrenocorticotropic hormone) or corticotropin is a 39-amino acid peptide hormone (MW=4500) secreted by the pituitary to regulate the production of steroid hormones by the adrenal cortex. ACTH secretion from the anterior pituitary is controlled by both a classical negative feedback control mechanism and CNS-stress mediated control system. Various types of stress or pain perceived in higher levels of the brain modulate secretion of the hypothalamic neurosecretory hormone, corticotropin releasing hormone (CRH), a 41-amino acid peptide. CRH stimulates pituitary ACTH secretion. The second peptide that modulates ACTH secretion is vasopressin (AVP). AVP secretion is also stimulated by stress and acts synergistically with CRH to increase ACTH secretion in the pituitary portal circulation. ACTH increases the synthesis and release of all adrenal sterioids, aldosterone, cortisol and adrenal androgens.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.