Rat CRF ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA835RA
MSPP-CEA835RAEA 684 USD
MSPP-CEA835RA
Rat CRF ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat CRF (Corticotropin Releasing Factor). The assay range is from 12.35 to 1000 pg/ml (Competitive kit) with a sensitivity of 5.53 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
releasing factor was initially isolated from ovine hypothalamus by Vale et al. in 1981, and identified as a novel neuropeptide comprising 41 amino acid residues with molecular weight 4758. Later human CRF and rat CRF were also isolated and identified. The mouse CRF peptide is identical at amino acid level to the rat and human CRF peptides. CRF in anterior pituitary promotes the synthesis and secretion of ACTH, a main factor of hypothalamus-pituitary-adrenal (HPA) axis. In the rat and human, CRF distributes mainly in hypothalamus, but it was also found in spinal cord, stomach, spleen, duodenum, adrenal and placenta. In addition, immunochemical evidence supported the wide distribution of the peptide throughout the central nervous system (CNS) such as olfactory bulb, retina and central auditory system in the rat.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.