Mouse vWF ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA833MU
MSPP-CEA833MUEA 648 USD
MSPP-CEA833MU
Mouse vWF ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse vWF (Von Willebrand Factor). The assay range is from 12.35 to 1,000 ng/ml (Competitive kit) with a sensitivity of 4.82 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Von Willebrand factor (vWF) is a blood glycoprotein involved in hemostasis. It is deficient or defective in von Willebrand disease and is involved in a large number of other diseases, including thrombotic thrombocytopenic purpura, Heyde's syndrome, and possibly hemolytic-uremic syndrome. It is a large multimeric glycoprotein present in blood plasma and produced constitutively in endothelium (in the Weibel-Palade bodies), megakaryocytes (α-granules of platelets), and subendothelial connective tissue. Besides, it is not an enzyme and therefore has no catalytic activity. Its primary function is binding to other proteins, particularly Factor VIII and it is important in platelet adhesion to wound sites.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.