Mouse FSH ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA830MU
MSPP-CEA830MUEA 648 USD
MSPP-CEA830MU
Mouse FSH ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse FSH (Follicle Stimulating Hormone). The assay range is from 2.22 to 180 ng/ml (Competitive kit) with a sensitivity of 0.77 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Follicle stimulating hormone (FSH) is a hormone synthesised and secreted by gonadotropes in the anterior pituitary gland. In the ovary FSH stimulates the growth of immature Graafian follicles to maturation. FSH is a glycoprotein secreted by the basophil cells of the anterior pituitary. Gonadotropin-releasing hormone (GnRH), produced in the hypothalamus, controls the release of FSH from the anterior pituitary. Like other glycoproteins, such as LH, TSH, and HCG, FSH consists of subunits designated as alpha and beta. Hormones of this type have alpha subunits that are very similar in structure, therefore the biological and immunological properties of each hormone are dependent on the unique beta subunit. In the female, FSH stimulates the growth and maturation of ovarian follicles by acting directly on the receptors located on the granulosa cells, follicular steroidogenesis is promoted and LH production is stimulated.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.