General Species SPH ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA805GE
MSPP-CEA805GEEA 796 USD
MSPP-CEA805GE
General Species SPH ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species SPH (Sphingomyelin). The assay range is from 1.23 to 100 ng/ml (Competitive kit) with a sensitivity of 0.55 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Sphingomyelin (SPH), is a type of sphingolipid found in animal cell membranes, especially in the membranous myelin sheath which surrounds some nerve cell axons. It usually consists of phosphorylcholine and ceramide. In humans SPH represents ~85% of all sphingolipids. In humans, sphingomyelin is believed to be the only cell membrane phospholipid not derived from glycerol. Like all sphingolipids, SPH has a ceramide core. In addition it contains one polar head group, which is either phosphocholine or phosphoethanolamine.The plasma membrane of cells is highly enriched in sphingomyelin and is considered largely to be found in the exoplasmic leaflet of the cell membrane. Neutral sphingomyelinase-2 an enzyme that breaks down sphingomyelin into ceramide has been found to localise exclusively to the inner leaflet further suggesting that there may be sphingomyelin present there.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.