General Species PGF2a ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA749GE
MSPP-CEA749GEEA 796 USD
MSPP-CEA749GE
General Species PGF2a ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species PGF2a (Prostaglandin F2 Alpha). The assay range is from 9.88 to 800 pg/ml (Competitive kit) with a sensitivity of 4.13 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
One of the prostaglandins, a group of hormone-like substances that participate in a wide range of body functions such as the contraction and relaxation of smooth muscle, the dilation and constriction of blood vessels, control of blood pressure, and modulation of inflammation. Prostaglandin F2-alpha(PGF-2 alpha) is a stable prostaglandin that stimulates the contraction of uterine and bronchial smooth muscle and produces vasoconstriction (tightening) in some blood vessels. As a pharmaceutical, the generic name of PGF-2 alpha is dinoprost. It is used for the induction of abortion, for evacuation of the uterus after a missed abortion, and in the treatment of hydatidiform mole.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.