General Species 8-EpiPGF2-α ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA701GE
MSPP-CEA701GEEA 796 USD
MSPP-CEA701GE
General Species 8-EpiPGF2-α ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species 8 to EpiPGF2 to α (8 to Epi Prostaglandin F2 Alpha). The assay range is from 24.69 to 2,000 pg/ml (Competitive kit) with a sensitivity of 8.73 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
The isoprostanes are a family of eicosanoids of non-enzymatic origin produced by the random oxidation of tissue phospholipids by oxygen radicals. Isoprostanes appear as artifacts in tissue and plasma samples which have undergone oxidative degradation during prolonged or improper storage. They also appear in the plasma and urine under normal conditions and are elevated by oxidative stress. At least one of the isoprostanes, 8-isoprostane (8-epi PGF2α), has been shown to have biological activity. It is a potent pulmonary and renal vasoconstrictor1 and has been implicated as a causative mediator of hepatorenal syndrome and pulmonary oxygen toxicity. 8-Isoprostane has been proposed as a marker of antioxidant deficiency and oxidative stress and elevated levels have been found in heavy smokers. 8-Isoprostane levels are also a relative indicator of sample integrity for lipid-containing samples such as serum, plasma, and whole cell preparations.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.