Chicken sIgA ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA641GA
MSPP-CEA641GAEA 684 USD
MSPP-CEA641GA
Chicken sIgA ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Chicken sIgA (Secretory Immunoglobulin A). The assay range is from 617.3 to 50,000 pg/ml (Competitive kit) with a sensitivity of 297.2 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Secretory IgA consists of two IgA monomers joined by the J-chain and furthermore a secretory component. It is secreted in plasma cells based in the lamina propia of mucosal membranes. Synthesis of IgA is independent from the synthesis of serum IgA. This means lack of serum IgA does not necessarily mean a lack of IgA. IgA is found in secretions in a specific form called secretory IgA, Secretory IgA is the major immunoglobulin in saliva, tears, colostrum, nasal mucous, mother's milk, tracheobronchial and gastrointestinal secretes. It plays a major role in preventing adherence of microorganisms to mucosal sites, in activation of the alternative complement pathway and in activating inflammatory reactions. Newborns are provided with IgA by mother's milk and are passively immunized against gastrointestinal infections.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.