General Species FA ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA610GE
MSPP-CEA610GEEA 796 USD
MSPP-CEA610GE
General Species FA ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species FA (Folic Acid). The assay range is from 781.25 to 200,000 pg/ml (Competitive kit) with a sensitivity of 285.77 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Folic acid is a form of the water-soluble vitamin B9. Folate is a naturally occurring form of the vitamin, found in food, while folic acid is synthetically produced, and used in fortified foods and supplements. Folic acid is itself not biologically active, but its biological importance is due to tetrahydrofolate and other derivatives after its conversion to dihydrofolic acid in the liver. Vitamin B9 (folic acid and folate) is essential for numerous bodily functions. Humans cannot synthesize folate de novo, therefore, folate has to be supplied through the diet to meet their daily requirements. The human body needs folate to synthesize DNA, repair DNA, and methylate DNA as well as to act as a cofactor in certain biological reactions. It is especially important in aiding rapid cell division and growth, such as in infancy and pregnancy. Children and adults both require folic acid to produce healthy red blood cells and prevent anemia.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.