General Species LTB4 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA562GE
MSPP-CEA562GEEA 796 USD
MSPP-CEA562GE
General Species LTB4 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species LTB4 (Leukotriene B4). The assay range is from 246.9 to 20000 pg/ml (Competitive kit) with a sensitivity of 90.1 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Leukotriene B4 is a leukotriene involved in inflammation. It is produced from leukocytes in response to inflammatory mediators and is able to induce the adhesion and activation of leukocytes on the endothelium, allowing them to bind to and cross it into the tissue. In neutrophils, it is also a potent chemoattractant, and is able to induce the formation of reactive oxygen species and the release of lysosome enzymes by these cells. Leukotriene B4 is a potent pro-inflammatory molecule that belongs to a family of eicosanoid lipid mediators. It is synthesized from arachidonic acid that is generated from nuclear membrane phospholipid. In general, and upon cell activation, phospholipase A2 is directed to the nuclear membrane. Here, it generates arachidonic acid that is bound by one 18 kDa membrane-bound protein termed Five-Lipooxygenase Activating Protein (FLAP).
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.