Human BNP ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA541HU
MSPP-CEA541HUEA 630 USD
MSPP-CEA541HU
Human BNP ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human BNP (Brain Natriuretic Peptide). The assay range is from 24.69 to 2000 pg/ml (Competitive kit) with a sensitivity of 9.67 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Brain natriuretic peptide is 32 amino acid polypeptide secreted by the ventricles of the heart in response to excessive stretching of heart muscle cells (cardiomyocytes). BNP is named as such because it was originally identified in extracts of porcine brain, although in humans it is produced mainly in the cardiac ventricles. BNP is co-secreted along with a 76 amino acid N-terminal fragment (NT-proBNP) which is biologically inactive. BNP binds to and activates the atrial natriuretic factor receptors NPRA, and to a lesser extent NPRB, in a fashion similar to atrial natriuretic peptide (ANP) but with 10-fold lower affinity. The biological half-life of BNP, however, is twice as long as that of ANP, and that of NT-proBNP is even longer, making these peptides better targets than ANP for diagnostic blood testing.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.