Mouse D2D ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA506MU
MSPP-CEA506MUEA 576 USD
MSPP-CEA506MU
Mouse D2D ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse D2D (D to Dimer). The assay range is from 617.3 to 50,000 pg/ml (Competitive kit) with a sensitivity of 243.7 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
D-dimers are unique in that they are the breakdown products of a fibrin mesh that has been stabilized by Factor XIII. This factor crosslinks the E-element to two D-elements. This is the final step in the generation of a thrombus. Plasmin is a fibrinolytic enzyme that organizes clots and breaks down the fibrin mesh. It cannot, however, break down the bonds between one E and two D units. The protein fragment thus left over is a D-dimer. D-dimer concentration may be determined by a blood test to help diagnose thrombosis. Since its introduction in the 1990s, it has become an important test performed in patients suspected of thrombotic disorders. While a negative result practically rules out thrombosis, a positive result can indicate thrombosis but does not rule out other potential causes. Its main use, therefore, is to exclude thromboembolic disease where the probability is low.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.