Human EDN1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA482HU
MSPP-CEA482HUEA 630 USD
MSPP-CEA482HU
Human EDN1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human EDN1 (Endothelin 1). The assay range is from 6.17 to 500 pg/ml (Competitive kit) with a sensitivity of 2.71 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Endothelin-1(ET-1), a peptide of 21 amino acid residues, is the most potent vasoconstrictive substance known. Originally isolated from porcine aortic endothelial cells, ET-1 is now known to be one of a family of three mammalian vasoactive peptides that also includes Endothelin-2 (ET-2) and Endothelin-3 (ET-3). These related peptides differ from ET-1 at two and six amino acid residue positions, respectively. A fourth peptide, vasoactive intestinal contractor (VIC), is sometimes classified as rat ET-2. All members of the endothelin family contain two essential disulfide bridges and six conserved amino acid residues at the C-terminus. Additionally, all of the endothelin family members are synthesized initially as prepropolypeptides of approximately 200 amino acid residues encoded by separate genes.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.