Rat CT ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA472RA
MSPP-CEA472RAEA 684 USD
MSPP-CEA472RA
Rat CT ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat CT (Calcitonin). The assay range is from 12.35 to 1000 pg/ml (Competitive kit) with a sensitivity of 4.95 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
The most prominent clinical syndrome associated with a disordered hypersecretion of Calcitonin is medullary carcinoma of the thyroid (MTC). MTC is a tumor of the Calcitonin producing C-cells of the thyroid gland. Although MTC is rare, comprising 5 to 10% of all thyroid cancer, it is often fatal. It may occur sporadically or in a familial form that is transmitted as an autosomal dominant trait. MTC has great clinical importance because of its familial distribution. Further, it leant itself to be diagnosed early by serum Calcitonin and total cure for early sub-clinical disease is possible1. This is frequently associated with other clinical features and it has good potential for cure with surgery. Although a rare tumor, it can occur in a familial pattern1 as a Type II multiple endocrine neoplasia.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.