Human PI ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA379HU
MSPP-CEA379HUEA 700 USD
MSPP-CEA379HU
Human PI ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human PI (Proinsulin). The assay range is from 12.35 to 1000 pg/ml (Competitive kit) with a sensitivity of 5.21 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Proinsulin is synthesized in the pancreatic beta cells as a 9390 mw polypeptide of 86 amino acids. Proinsulin is subsequently cleaved enzymatically, releasing insulin into the circulation along with a residual 3000 mw fragment called C-peptide, so-named because it connects the A and B chains of insulin within the proinsulin molecule. Proinsulin, which has relatively low biological activity (approximately 10% of insulin potency), is the major storage form of insulin. Normally, only small amounts (~3% of the amount of insulin, on a molar basis) of proinsulin enter the circulation. Because the hepatic clearance of proinsulin is only 25% of insulin clearance, the half-life of proinsulin is two- to threefold longer and concentrations in the fasting state are approximately 10 to 15% of insulin concentrations.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.