Rat HbA1c ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA190RA
MSPP-CEA190RAEA 684 USD
MSPP-CEA190RA
Rat HbA1c ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat HbA1c (Glycated Hemoglobin A1c). The assay range is from 12.35-1000ug/mL (Competitive kit) with a sensitivity of 5.05ug/mL. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Hemoglobin A1c was first separated from other forms of hemoglobin by Huisman and Meyering in 1958 using a chromatographic column. In the blood stream are the red blood cells, which are made of a molecule, haemoglobin. Glucose sticks to the haemoglobin to make a 'glycosylated haemoglobin' molecule, called haemoglobin A1C or HbA1C. The more glucose in the blood, the more haemoglobin A1C or HbA1C will be present in the blood.  HbA1c is a form of hemoglobin used primarily to identify the average plasma glucose concentration over prolonged periods of time. It is formed in a non-enzymatic pathway by hemoglobin's normal exposure to high plasma levels of glucose. Glycation of hemoglobin has been associated with cardiovascular disease, nephropathy, and retinopathy in diabetes mellitus. Monitoring the HbA1c in type-1 diabetic patients may improve treatment.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.