General Species fT3 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA186GE
MSPP-CEA186GEEA 612 USD
MSPP-CEA186GE
General Species fT3 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species fT3 (Free Triiodothyronine). The assay range is from 740.7 to 60000fg/mL (Competitive kit) with a sensitivity of 249.5fg/mL. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Triiodothyronine (T3) is a thyroid hormone that circulates in blood almost completely bound (]99.5%) to carrier proteins. The main transport protein is thyroxine-binding globulin (TBG). Furthermore, the concentrations of the carrier proteins are altered in many clinical conditions, such as pregnancy. In normal thyroid function, as the concentrations of the carrier proteins changes, the total triiodothyronine level also changes, so that the free triiodothyronine concentration remains constant. Measurements of free triiodothyronine (Free T3) concentrations, therefore, correlate more reliably with your clinical status than total triiodothyronine (T3) levels. For example, the increase in total triiodothyronine levels associated with pregnancy, oral contraceptives and estrogen therapy result in higher total T3 levels while the free T3 concentration remains unchanged.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.