Mouse AngII ELISA Kit
Supplier: CLOUD-CLONE CORP MS
CEA005MU
MSPP-CEA005MUEA 648 USD
MSPP-CEA005MU
Mouse AngII ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse AngII (Angiotensin II). The assay range is from 24.69 to 2000 pg/ml (Competitive kit) with a sensitivity of 8.84 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Angiotensin I is converted to angiotensin II through removal of two terminal residues by the enzyme Angiotensin-converting enzyme (ACE, or kinase), which is found predominantly in the capillaries of the lung. ACE is actually found all over the body, but has its highest density in the lung due to the high density of capillary beds there. Angiotensin II acts as an endocrine, autocrine/paracrine, and intracrine hormone.ACE is a target for inactivation by ACE inhibitor drugs, which decrease the rate of angiotensin II production. Angiotensin II increases blood pressure by stimulating the Gq protein in vascular smooth muscle cells. ACE inhibitor drugs are major drugs against hypertension.Other cleavage products of ACE, 7 or 9 amino acids long, are also known; they have differential affinity for angiotensin receptors, although their exact role is still unclear.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.