Adapter, PTFE, Screw Thread, Analytical Sales and Services
Supplier: ANALYTICAL SALES & SERVICE MS
CA4505
MSPP-CA4505EA 138.46 USD
MSPP-CA4505
Adapter, PTFE, Screw Thread, Analytical Sales and Services
Fittings and Connectors
Fits GL45 thread safety caps, GL38 bottle and Canary-Safe™ caps.
Fits to most common 4 L solvent bottles.