OrbiShaker MP Microplate Shaker or Vortexer with Platform for Four Microplates, Southern Labware
Supplier: SOUTHERN LABWARE, INC MS
BT1502
MSPP-BT1502EA 1528.46 USD
MSPP-BT1502
OrbiShaker MP Microplate Shaker or Vortexer with Platform for Four Microplates, Southern Labware
Shakers and Mixers Microplate Shakers
The OrbiShaker MP is designed with a 3 mm mixing orbit, optimized for thorough mixing of microplates and/or PCR plates. Up to four plates (standard or deep well) can be placed on the included platform and are instantly secured without the use of springs, clamps or tools.
  • Digital speed control up to 1500 rpm
  • 3 mm orbit for thorough mixing in microplates
  • Includes platform for up to four microplates
  • Cold room and incubator safe
  • Optional adapters for mixing microtubes